Isopropyl myristate ≥98%

Synonyms: Tetradecanoic acid isopropyl ester, Myristic acid isopropyl ester, Isopropyl Myristate, Isopropyl tetradecanoate, Tetradecylic acid isopropyl ester

172472-1L 172472-25ML
220039-431EA 211.08 USD
220039-431 220039-432
Isopropyl myristate ≥98%
Isopropyl myristate
Formula: C₁₇H₃₄O₂
MW: 270.46 g/mol
Boiling Pt: 320 °C (1013 hPa)
Melting Pt: 0…1 °C
Density: 0.852 g/cm³ (20 °C)
Flash Pt: 150 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008982
CAS Number: 110-27-0
EINECS: 203-751-4
Merck Index: 13,05235

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR