1-Iodopropane ≥99%

Danger

Synonyms: 1-Propyl iodide

171883-100G 171883-500G 171883-5G
220039-835EA 52.98 USD
220039-835 220039-836 220039-837
1-Iodopropane ≥99%
1-Iodopropane
Formula: C₃H₇I
MW: 169.99 g/mol
Boiling Pt: 103 °C (1013 hPa)
Melting Pt: –98 °C
Density: 1.741 g/cm³ (20 °C)
Flash Pt: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD00001095
CAS Number: 107-08-4
EINECS: 203-460-2
UN: 2392
ADR: 3,III
Merck Index: 12,08047

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR