Methyl propiolate ≥99%

Synonyms: Methyl acetylenecarboxylate, Propiolic acid methyl ester, 2-Propynoic acid methyl ester

171859-10G 171859-2.5G 171859-50G
220039-831EA 114.15 USD
220039-831 220039-832 220039-833
Methyl propiolate ≥99%
Methyl propiolate
Formula: C₄H₄O₂
MW: 84.07 g/mol
Boiling Pt: 105 °C (1013 hPa)
Density: 1.196 g/cm³ (20 °C)
MDL Number: MFCD00008572
CAS Number: 922-67-8
EINECS: 213-083-5
UN: 3272
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR