DL-Proline

Synonyms: (±)-Proline, H-DL-Pro-OH

171824-5G 171824-1G
220039-828EA 46.42 USD
220039-828 220039-827
DL-Proline
DL-Proline
Formula: C₅H₉NO₂
MW: 115.13 g/mol
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 208…210 °C
Density: 1.38 g/cm³ (22.9 °C)
MDL Number: MFCD00005250
CAS Number: 609-36-9
EINECS: 210-189-3
Merck Index: 12,07963

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR