2-Aminobenzimidazole

Synonyms: 1H-Benzo[d]imidazol-2-amine, Benzimidazol-2-ylamine, 2-Amino-1H-benzimidazole

171778-5G 171778-25G
220038-494EA 38.66 USD
220038-494 220038-493
2-Aminobenzimidazole
2-Aminobenzimidazole
Formula: C₇H₇N₃
MW: 133.15 g/mol
Boiling Pt: 381 °C (1013 hPa)
Melting Pt: 226…231 °C
MDL Number: MFCD00005596
CAS Number: 934-32-7
EINECS: 213-280-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR