Undecanoic acid ≥98%

Synonyms: Hendecanoic acid, Undecylic acid

171476-100G 171476-25G
220038-487EA 49.54 USD
220038-487 220038-488
Undecanoic acid ≥98%
Undecanoic acid
Formula: C₁₁H₂₂O₂
MW: 186.29 g/mol
Boiling Pt: 228 °C (1013 hPa)
Melting Pt: 30 °C
Density: 0.89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002730
CAS Number: 112-37-8
EINECS: 203-964-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR