Pentadecafluorocaprylic acid ≥96%

Synonyms: Pentadecafluorooctoic acid, Pentadecafluorooctanoic acid, PFOA, Perfluorocaprylic acid, Perfluorooctanoic acid

171468-5G 171468-25G
220039-824EA 45.12 USD
220039-824 220039-823
Pentadecafluorocaprylic acid ≥96%
Pentadecafluorocaprylic acid
Formula: C₈HF₁₅O₂
MW: 414.07 g/mol
MDL Number: MFCD00004174
CAS Number: 335-67-1
EINECS: 206-397-9
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR