Stearoyl chloride ≥97%

171158-5G 171158-25G
220039-801EA 130.31 USD
220039-801 220039-800
Stearoyl chloride ≥97%
Stearoyl chloride
Formula: C₁₈H₃₅ClO
MW: 302.93 g/mol
Boiling Pt: 348 °C (1013 hPa)
Melting Pt: 164…167 °C
Density: 0.9007 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000744
CAS Number: 112-76-5
EINECS: 204-004-5
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR