5-Nitro-2-furaldehyde

Synonyms: 5-Nitrofurfural

170968-25G 170968-5G
220039-788EA 169.08 USD
220039-788 220039-789
5-Nitro-2-furaldehyde
5-Nitro-2-furaldehyde
Formula: C₅H₃NO₄
MW: 141.08 g/mol
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 35 °C
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003230
CAS Number: 698-63-5
EINECS: 211-816-3
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR