(4-Methoxyphenyl)acetonitrile ≥97%

Synonyms: 4-Methoxybenzyl cyanide, 4-Methoxyphenylacetonitrile

169986-100G 169986-25G
220039-734EA 100.48 USD
220039-734 220039-735
(4-Methoxyphenyl)acetonitrile ≥97%
(4-Methoxyphenyl)acetonitrile
Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 286 °C (1013 hPa)
Melting Pt: 8 °C
Density: 1.085 g/cm³ (20 °C)
Flash Pt: 117 °C
MDL Number: MFCD00001919
CAS Number: 104-47-2
EINECS: 203-206-0
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR