Diphenyl disulfide ≥99%

Synonyms: 1,2-Diphenyldisulphane, Phenyl disulphide

169021-10G 169021-50G 169021-250G
220039-649EA 28.86 USD
220039-649 220039-651 220039-650
Diphenyl disulfide ≥99%
Diphenyl disulphide
Formula: C₁₂H₁₀S₂
MW: 218.34 g/mol
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 58…60 °C
Density: 1.155 g/cm³ (64.9 °C)
MDL Number: MFCD00003065
CAS Number: 882-33-7
EINECS: 212-926-4
UN: 3335
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR