4,4'-Biphenol ≥97%

Synonyms: [1,1'-Biphenyl]-4,4'-diol, 4,4'-Dihydroxybiphenyl, Biphenyl-4,4'-diol

168734-25G 168734-100G
220039-576EA 149.08 USD
220039-576 220039-575
4,4'-Biphenol ≥97%
4,4'-Biphenol
Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
Boiling Pt: 355 °C (1013 hPa)
Density: 1.25 g/cm³ (20 °C)
MDL Number: MFCD00002348
CAS Number: 92-88-6
EINECS: 202-200-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR