2,4-Dihydroxybenzaldehyde ≥98%

Warning

168637-25G 168637-100G
220038-917EA 70.54 USD
220038-917 220038-986
2,4-Dihydroxybenzaldehyde ≥98%
2,4-Dihydroxybenzaldehyde
Formula: C₇H₆O₃
MW: 138.12 g/mol
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 132…133 °C
Density: 1.448 g/cm³ (-93 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011686
CAS Number: 95-01-2
EINECS: 202-383-1
Merck Index: 12,08324

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR