Di-tert-butyl peroxide ≥98%

Danger

Synonyms: Luperox(R) DI, tert-Butyl peroxide, tert-Butyl peroxide

168521-1L 168521-5ML 168521-250ML
220038-980EA 126.69 USD
220038-980 220038-982 220038-981
Di-tert-butyl peroxide ≥98%
Di-tert-butyl peroxide
Formula: C₈H₁₈O₂
MW: 146.23 g/mol
Boiling Pt: 109 °C (1013 hPa)
Melting Pt: < –25 °C
Density: 0.798 g/cm³ (20 °C)
Flash Pt: 12 °C
MDL Number: MFCD00008803
CAS Number: 110-05-4
EINECS: 203-733-6
Merck Index: 12,03515

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR