L(+)-Cysteine ≥97%

Warning

Synonyms: H-Cys-OH, (R)-(+)-Cysteine, (R)-2-Amino-3-mercaptopropanoic acid

168149-100G 168149-2.5G 168149-500G 168149-25G
220039-570EA 83.89 USD
220039-570 220039-571 220039-573 220039-572
L(+)-Cysteine ≥97%
L(+)-Cysteine
Formula: HSCH₂CH(NH₂)CO₂H
MW: 121.16 g/mol
Boiling Pt: 254 °C (1013 hPa)
Melting Pt: 115 °C
Density: 1.523 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064306
CAS Number: 52-90-4
EINECS: 200-158-2

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR