4-Bromo-1-butene ≥97%

167851-1G 167851-50G 167851-10G
220039-490EA 30.91 USD
220039-490 220039-491 220038-839
4-Bromo-1-butene ≥97%
4-Bromo-1-butene
Formula: C₄H₇Br
MW: 135 g/mol
Boiling Pt: 98 °C (1013 hPa)
Density: 1.33 g/cm³ (20 °C)
Flash Pt: 9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000258
CAS Number: 5162-44-7
EINECS: 225-937-4
UN: 1992
ADR: 3,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR