α'-Chloro-α,α,α-trifluoro-m-xylene ≥97%

Synonyms: 3-(Trifluoromethyl)benzyl chloride, 3-(Chloromethyl)benzotrifluoride, 1-(Chloromethyl)-3-(trifluoromethyl)benzene

167703-5G 167703-25G
220038-835EA 30.26 USD
220038-835 220038-834
α'-Chloro-α,α,α-trifluoro-m-xylene ≥97%
α'-Chloro-α,α,α-trifluoro-m-xylene
Formula: C₈H₆ClF₃
MW: 194.58 g/mol
Boiling Pt: 95 °C (1013 hPa)
Density: 1.3079 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000908
CAS Number: 705-29-3
EINECS: 211-884-4
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR