2-Chlorobenzothiazole

167576-25G 167576-5G
220038-825EA 164.77 USD
220038-825 220038-826
2-Chlorobenzothiazole
2-Chlorbenzothiazol
Formula: C₇H₄ClNS
MW: 169.63 g/mol
Boiling Pt: 141 °C (1013 hPa)
Melting Pt: 21…24 °C
Density: 1.37 g/cm³ (20 °C)
MDL Number: MFCD00005776
CAS Number: 615-20-3
EINECS: 210-415-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR