Chloroacetone ≥95%

Danger

Synonyms: Chloropropanone

167479-500G 167479-100G 167479-5G
220039-488EA 80.77 USD
220039-488 220039-487 220039-489
Chloroacetone ≥95%
Chloroacetone
Formula: C₃H₅ClO
MW: 92.52 g/mol
Boiling Pt: 120 °C (1013 hPa)
Melting Pt: -44 °C
Density: 1.15 g/cm³ (20 °C)
Flash Pt: 36 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000936
CAS Number: 78-95-5
EINECS: 201-161-1
UN: 1695
ADR: 6.1,I
Merck Index: 13,02131

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR