tert-Butyl carbamate ≥98%

Synonyms: Carbamic acid tert-butyl ester

167398-1G 167398-5G
220039-485EA 247.69 USD
220039-485 220039-486
tert-Butyl carbamate ≥98%
tert-Butyl carbamate
Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
MDL Number: MFCD00007962
CAS Number: 4248-19-5
EINECS: 224-209-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR