1-Iodobutane ≥99%

Danger

Synonyms: 1-Butyl iodide

167304-500G 167304-100G 167304-5G
220039-482EA 144.31 USD
220039-482 220039-481 220039-483
1-Iodobutane ≥99%
1-Iodobutane
Formula: C₄H₉I
MW: 184.02 g/mol
Boiling Pt: 130 °C (1013 hPa)
Melting Pt: -103 °C
Density: 1.616 g/cm³ (20 °C)
Flash Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00001098
CAS Number: 542-69-8
EINECS: 208-824-4
UN: 1993
ADR: 3,III
Merck Index: 13,01572

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR