2-Bromo-p-xylene ≥99%

Warning

Synonyms: 2-Bromo-1,4-dimethylbenzene, 2,5-Dimethylbromobenzene

167266-25G 167266-100G
220039-479EA 31.23 USD
220039-479 220039-478
2-Bromo-p-xylene ≥99%
2-Bromo-p-xylene
Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: 8.9 °C
Density: 1.3582 g/cm³ (18.5 °C)
MDL Number: MFCD00000074
CAS Number: 553-94-6
EINECS: 209-055-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR