3-Bromo-1-propanol ≥97%

Warning

Synonyms: 3-Bromopropan-1-ol, 1-Bromo-3-hydroxypropane

167169-100G 167169-5G 167169-25G
220039-465EA 465.23 USD
220039-465 220039-467 220039-466
3-Bromo-1-propanol ≥97%
3-Bromo-1-propanol
Formula: BrH₂CCH₂CH₂OH
MW: 138.99 g/mol
Boiling Pt: 145 °C (1013 hPa)
Density: 1.6 g/cm³ (20 °C)
Flash Pt: 65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002942
CAS Number: 627-18-9
EINECS: 210-986-6

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR