1-Bromo-4-nitrobenzene ≥99%

Synonyms: 4-Bromonitrobenzene

167150-5G 167150-25G
220039-419EA 30.37 USD
220039-419 220039-418
1-Bromo-4-nitrobenzene ≥99%
1-Bromo-4-nitrobenzene
Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: 124…126 °C
Density: 1.93 g/cm³ (20 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007280
CAS Number: 586-78-7
EINECS: 209-583-8
UN: 3459
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR