11-Bromoundecanoic acid ≥95%, Technical Grade

Synonyms: 11-Bromohendecanoic acid, 11-Bromoundecylic acid

165816-1KG 165816-250G
220039-371EA 777.54 USD
220039-371 220039-372
11-Bromoundecanoic acid ≥95%, Technical Grade
11-Bromoundecanoic acid
Formula: BrH₂C(CH₂)₉COOH
MW: 265.19 g/mol
Boiling Pt: 354 °C (1013 hPa)
Melting Pt: 48…49 °C
Density: 1.24 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002732
CAS Number: 2834-05-1
EINECS: 220-602-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR