Methylboronic acid ≥97%

165336-1G 165336-5G
220039-348EA 36.42 USD
220039-348 220039-349
Methylboronic acid ≥97%
Methylboronic acid
Formula: CH₅BO₂
MW: 59.86 g/mol
MDL Number: MFCD00002105
CAS Number: 13061-96-6

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR