Adipoyl dichloride ≥98%

Danger

Synonyms: Adipoyl chloride

165212-25G 165212-5G 165212-100G
220038-682EA 91.8 USD
220038-682 220038-683 220038-681
Adipoyl dichloride ≥98%
Adipoyl dichloride
Formula: C₆H₈Cl₂O₂
MW: 183.03 g/mol
Boiling Pt: 125 °C (15 hPa)
Density: 1.2563 g/cm³ (20 °C)
Flash Pt: 66 °C
MDL Number: MFCD00000759
CAS Number: 111-50-2
EINECS: 203-876-4
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR