o-Acetoxybenzoyl chloride ≥95%

Danger

Synonyms: 2-(Chlorocarbonyl)phenyl acetate, O-Acetylsalicyloyl chloride, 2-Acetylsalicyloyl chloride, 2-Acetoxybenzoyl chloride

165190-25G 165190-5G
220038-678EA 84.11 USD
220038-678 220038-679
o-Acetoxybenzoyl chloride ≥95%
o-Acetoxybenzoyl chloride
Formula: C₉H₇ClO₃
MW: 198.61 g/mol
Boiling Pt: 346 °C (1013 hPa)
Melting Pt: 45 - 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00000663
CAS Number: 5538-51-2
EINECS: 226-899-1
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR