3-Oxoglutaric acid, Technical Grade

Synonyms: Acetone-1,3-dicarboxylic acid, 3-Ketoglutaric acid, 3-Oxopentanedioic acid, 1,3-Acetonedicarboxylic acid, β-Ketoglutaric acid

165115-25G 165115-100G
220038-669EA 52.69 USD
220038-669 220038-668
3-Oxoglutaric acid, Technical Grade
3-Oxoglutaric acid
Formula: C₅H₆O₅
MW: 146.1 g/mol
Boiling Pt: 323 °C (1013 hPa)
Melting Pt: 138 °C
Density: 1.55 g/cm³ (20 °C)
Storage Temperature: Freezer
MDL Number: MFCD00002711
CAS Number: 542-05-2
EINECS: 208-797-9
Merck Index: 13,00069

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR