Tritylium tetrafluoroborate

Synonyms: Triphenylcarbenium tetrafluoroborate

164577-25G 164577-5G
220038-969EA 138.46 USD
220038-969 220038-970
Tritylium tetrafluoroborate
Tritylium tetrafluoroborate
Formula: C₁₉H₁₅BF₄
MW: 330.13 g/mol
Melting Pt: 212 °C
MDL Number: MFCD00013120
CAS Number: 341-02-6
EINECS: 206-433-3
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR