3-Amino-2-naphthol

Synonyms: 3-Amino-2-hydroxynaphthalene, 3-Aminonaphthalen-2-ol

164267-5G 164267-1G
220039-321EA 190.38 USD
220039-321 220039-320
3-Amino-2-naphthol
3-Amino-2-naphthol
Formula: C₁₀H₉NO
MW: 159.19 g/mol
MDL Number: MFCD00004113
CAS Number: 5417-63-0
EINECS: 226-519-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR