3-Methoxy-2-nitrobenzaldehyde ≥97%

163821-25G 163821-5G
220038-886EA 764.62 USD
220038-886 220038-887
3-Methoxy-2-nitrobenzaldehyde ≥97%
3-Methoxy-2-nitrobenzaldehyde
Formula: C₈H₇NO₄
MW: 181.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007135
CAS Number: 53055-05-3
EINECS: 258-332-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR