Bromobenzene

Danger

Synonyms: Phenylbromide, Phenyl bromide

16350-100ML 16350-1L 16350-500ML
220039-290EA 45.23 USD
220039-290 220039-291 220039-292
Bromobenzene
Bromobenzene
Formula: C₆H₅Br
MW: 157.01 g/mol
Boiling Pt: 155 °C (1013 hPa)
Melting Pt: –31 °C
Density: 1.495 g/cm³ (20 °C)
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00000055
CAS Number: 108-86-1
EINECS: 203-623-8
UN: 2514
ADR: 3,III
Merck Index: 13,01390

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR