2,6-Di-tert-butyl-4-methoxyphenol ≥98%

Synonyms: 3,5-Di-tert-butyl-4-hydroxyanisole

163392-5G 163392-25G
220039-284EA 120.62 USD
220039-284 220039-283
2,6-Di-tert-butyl-4-methoxyphenol ≥98%
2,6-Di-tert-butyl-4-methoxyphenol
Formula: C₁₅H₂₄O₂
MW: 236.35 g/mol
Boiling Pt: 135…140 °C (10 mmHg)
Melting Pt: 105.4 °C
MDL Number: MFCD00008824
CAS Number: 489-01-0
EINECS: 207-693-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR