Geraniol

Warning

Synonyms: (E)-3,7-Dimethylocta-2,6-dien-1-ol

163333-25G 163333-100G
220039-275EA 49.97 USD
220039-275 220039-274
Geraniol
Geraniol
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
MDL Number: MFCD00002917
CAS Number: 106-24-1
EINECS: 203-377-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR