1-Butylboronic acid ≥97%

Synonyms: Butylboronic acid, n-Butylboronic acid

163244-5G 163244-1G 163244-25G
220038-809EA 75.46 USD
220038-809 220038-807 220038-808
1-Butylboronic acid ≥97%
1-Butylboronic acid
Formula: C₄H₁₁BO₂
MW: 101.94 g/mol
MDL Number: MFCD00002106
CAS Number: 4426-47-5
EINECS: 224-607-7

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR