Thallium(III) nitrate trihydrate

Synonyms: Thallium trinitrate trihydrate

163015-100G 163015-25G
220038-552EA 726.92 USD
220038-552 220038-553
Thallium(III) nitrate trihydrate
Thallium(III) nitrate trihydrate
Formula: N₃O₉Tl·3H₂O
MW: 444.44 g/mol
Melting Pt: 102…105 °C
Density: 3.31 to 3.37 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149874
CAS Number: 13453-38-8
EINECS: 237-325-4
UN: 2727
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR