2-(2-Chloroethoxy)ethanol ≥99%

Warning

162965-250G 162965-50G
220039-272EA 172.31 USD
220039-272 220039-273
2-(2-Chloroethoxy)ethanol ≥99%
2-(2-Chloroethoxy)ethanol
Formula: C₄H₉ClO₂
MW: 124.57 g/mol
Boiling Pt: 79 °C (1013 hPa)
Density: 1.175 g/cm³ (20 °C)
MDL Number: MFCD00002870
CAS Number: 628-89-7
EINECS: 211-059-9
UN: 3334
ADR: 9,

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR