(±)-2-Aminobutyric acid ≥99%

162663-100G 162663-25G
220039-260EA 149.69 USD
220039-260 220039-261
(±)-2-Aminobutyric acid ≥99%
(±)-2-Aminobutyric acid
Formula: C₄H₉NO₂
MW: 103.12 g/mol
MDL Number: MFCD00008093
CAS Number: 2835-81-6
EINECS: 220-616-5

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR