2-Amino-6-methoxybenzothiazole

Synonyms: 6-Methoxybenzothiazol-2-ylamine

162590-50G 162590-250G
220039-255EA 154.62 USD
220039-255 220039-254
2-Amino-6-methoxybenzothiazole
2-Amino-6-methoxybenzothiazole
Formula: C₈H₈N₂OS
MW: 180.23 g/mol
Boiling Pt: 348 °C (1013 hPa)
Melting Pt: 160 °C
MDL Number: MFCD00005787
CAS Number: 1747-60-0
EINECS: 217-130-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR