Dimethyl glyoxime ≥99% ACS

Danger

Synonyms: Diacetyl glyoxime, Diacetyl dioxime, 2,3-Butanedione dioxime

162574-500G 162574-100G
220039-251EA 160 USD
220039-251 220039-250
Dimethyl glyoxime ≥99% ACS
Dimethyl glyoxime
Formula: CH₃C(=NOH)C(=NOH)CH₃
MW: 116.12 g/mol
Boiling Pt: 53 °C (8 mmHg)
Melting Pt: 240…241 °C
Density: 1.37 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002117
CAS Number: 95-45-4
EINECS: 202-420-1
Merck Index: 13,03272

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR