3-Fluorobenzoyl chloride ≥98%

Danger

Synonyms: 3-Fluorobenzoylchloride, m-Fluorobenzoyl chloride

162531-25G 162531-5G
220039-243EA 463.08 USD
220039-243 220039-244
3-Fluorobenzoyl chloride ≥98%
3-Fluorobenzoyl chloride
Formula: C₇H₄ClFO
MW: 158.56 g/mol
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: –30 °C
Density: 1.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000670
CAS Number: 1711-07-5
EINECS: 216-977-3
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR