3-Methyl-2-buten-1-ol ≥99%

Synonyms: 2-Methyl-2-buten-4-ol

162353-5ML 162353-100ML 162353-500ML
220039-234EA 37.8 USD
220039-234 220039-232 220039-233
3-Methyl-2-buten-1-ol ≥99%
3-Methyl-2-buten-1-ol
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: –86 °C
Density: 0.848 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002916
CAS Number: 556-82-1
EINECS: 209-141-4
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR