1-Chloro-3-fluorobenzene ≥99%

162299-25G 162299-5G
220039-222EA 103.92 USD
220039-222 220039-223
1-Chloro-3-fluorobenzene ≥99%
1-Chloro-3-fluorobenzene
Formula: C₆H₄ClF
MW: 130.55 g/mol
Boiling Pt: 126 °C (1013 hPa)
Density: 1.221 g/cm³ (25 °C)
MDL Number: MFCD00000569
CAS Number: 625-98-9
EINECS: 210-919-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR