1-Phenyl-1-propyne ≥99%

Warning

Synonyms: 1-Propynylbenzene, Prop-1-yn-1-ylbenzene

161241-25G 161241-5G
220039-119EA 372.62 USD
220039-119 220039-120
1-Phenyl-1-propyne ≥99%
1-Phenyl-1-propyne
Formula: C₆H₅CCCH₃
MW: 116.16 g/mol
Boiling Pt: 185 °C (1013 hPa)
Density: 0.942 g/cm³ (15 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00009272
CAS Number: 673-32-5
EINECS: 211-607-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR