Bromoacetyl chloride ≥95% (by GC)

16120-100ML 16120-25ML 16120-500ML
220039-115EA 341.38 USD
220039-115 220039-116 220039-117
Bromoacetyl chloride ≥95% (by GC)
Bromoacetyl chloride
Formula: BrCH₂COOCl
MW: 157.39 g/mol
MDL Number: MFCD00000724
CAS Number: 22118-09-8
EINECS: 244-790-7
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Assay 95% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR