5-Nitro-o-anisidine ≥98%

Synonyms: 2-Amino-4-nitroanisole, 2-Methoxy-5-nitroaniline

161195-100G 161195-500G
220039-113EA 2382.77 USD
220039-113 220039-114
5-Nitro-o-anisidine ≥98%
5-Nitro-o-anisidine
Formula: C₇H₈N₂O₃
MW: 168.15 g/mol
Boiling Pt: 352 °C (1013 hPa)
Melting Pt: 118 °C
Density: 1.2068 g/cm³ (156 °C)
MDL Number: MFCD00007261
CAS Number: 99-59-2
EINECS: 202-770-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR