meso-Tetraphenylporphine

Synonyms: 5,10,15,20-Tetraphenyl-21H,23H-porphine, 5,10,15,20-Tetraphenylporphyrin, meso-Tetraphenylporphine, TPP

160997-5G 160997-25G 160997-1G
220038-098EA 361.2 USD
220038-098 220038-097 220038-096
meso-Tetraphenylporphine
meso-Tetraphenylporphine
Formula: C₄₄H₃₀N₄
MW: 614.75 g/mol
Density: 1.276 g/cm³ (20 °C)
MDL Number: MFCD00011680
CAS Number: 917-23-7
EINECS: 213-025-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7bbe135263037ea3b97779308fb40e99ebbf19ce&vid=9bc20155ac97e4fe5383462bc7ed4796d57798b2&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR