2-Acetyl-1-methyl-1H-pyrrole

Synonyms: 1-(1-Methyl-1H-pyrrol-2-yl)ethanone, 2-Acetyl-1-methylpyrrole

160865-1G 160865-10G
220038-867EA 31.02 USD
220038-867 220038-866
2-Acetyl-1-methyl-1H-pyrrole
2-Acetyl-1-methyl-1H-pyrrole
Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 200 °C (1013 hPa)
Density: 1.0445 g/cm³ (15 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00003089
CAS Number: 932-16-1
EINECS: 213-247-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR