2-Nitro-m-cresol ≥99%

Synonyms: 2-Nitro-3-methylphenol, 3-Methyl-2-nitrophenol

160741-10G 160741-50G
220039-107EA 519.88 USD
220039-107 220039-108
2-Nitro-m-cresol ≥99%
2-Nitro-m-cresol
Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 231 °C (1013 hPa)
Melting Pt: 35…39 °C
MDL Number: MFCD00007125
CAS Number: 4920-77-8
EINECS: 225-546-9
UN: 2446
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR